Cerium(IV) oxide, REacton™, 99.9% (REO)
Cerium(IV) oxide, REacton™, 99.9% (REO)
Cerium(IV) oxide, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Cerium(IV) oxide, REacton™, 99.9% (REO)

CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 011328.30
also known as 011328-30
Price (USD)
137.65
Special offer
Online exclusive
Ends: 30-Jun-2026
162.00
Save 24.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
137.65
Special offer
Online exclusive
Ends: 30-Jun-2026
162.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS1306-38-3
IUPAC Namedioxocerium
Molecular FormulaCeO2
InChI KeyCETPSERCERDGAM-UHFFFAOYSA-N
SMILESO=[Ce]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream to brown
Total Rare Earth Oxide Impurities≤0.1%
Cerium(IV) oxide is used in ceramics, and to polish glass and stones. It is applied as a solid electrolyte in fuel cells. It can also be employed as a co-catalyst in a number of reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only