Cobalt(II) nitrate hexahydrate, 97.7% min
Cobalt(II) nitrate hexahydrate, 97.7% min
Cobalt(II) nitrate hexahydrate, 97.7% min
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, 97.7% min

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
5 kg
Catalog number 011341.A1
also known as 011341-A1
Price (USD)
269.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
269.00
Each
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.7% min
Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only