Cobalt(II) carbonate, 99% (metals basis), Co 45% min
Cobalt(II) carbonate, 99% (metals basis), Co 45% min
Cobalt(II) carbonate, 99% (metals basis), Co 45% min
Thermo Scientific Chemicals

Cobalt(II) carbonate, 99% (metals basis), Co 45% min

For Use in Ceramics, Manufacturing Co Compounds, Pigments | CAS: 513-79-1 | CCoO3 | 118.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 011343.36
also known as 011343-36
Price (USD)
225.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
225.00
Each
Chemical Identifiers
CAS513-79-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to Purple
Assay (unspecified)Co 45% min
Total Metal Impurities1% max.
Formpowder
Cobalt(II) carbonate is precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and also used in the manufacture of paints and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only