Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)
Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)
Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)
Thermo Scientific Chemicals

Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)

CAS: 7439-92-1 | Pb | 207.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
210 g
4 x 210 g
Catalog number 011346.18
also known as 011346-18
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
96.65
Online Exclusive
107.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0002% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only