Lithium thiocyanate hydrate
Lithium thiocyanate hydrate
Lithium thiocyanate hydrate
Thermo Scientific Chemicals

Lithium thiocyanate hydrate

CAS: 123333-85-7 | CLiNS | 65.02 g/mol
Have Questions?
Catalog NumberQuantity
011349.18
also known as 011349-18
50 g
Catalog number 011349.18
also known as 011349-18
Price (USD)
159.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
159.00
Each
Chemical Identifiers
CAS123333-85-7
IUPAC Namelithium(1+) cyanosulfanide
Molecular FormulaCLiNS
InChI KeyZJZXSOKJEJFHCP-UHFFFAOYSA-M
SMILES[Li+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormColorless crystalline mass
Assay (Argentometric Titration)LiSCN 63% min
Lithium thiocyanate hydrate is used as solution in infra red ultrasonic technique and also used as solvent for the fibers of mulberry in silk production process. It is used to determined the molecular structure of benzo-13-crown-4 compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only