Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
Thermo Scientific Chemicals

Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)

Form: Approx. 2.7g/100 x 100mm; Temper: as rolled | CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
100 x 400 mm
5 x 100 x 400 mm
Catalog number 011376.GH
also known as 011376-GH
Price (USD)
87.65
Online Exclusive
96.80
Save 9.15 (9%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
96.80
Save 9.15 (9%)
Each
Specifications
AppearanceSilver-colored foil
Assay Percent Range99.997% (Metals basis)
CAS7429-90-5
ChEBICHEBI:33629
Chemical Name or MaterialAluminum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only