Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Iron foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)

Form: approx. 0.49g/25x25mm | CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011381.FI
also known as 011381-FI
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormFoil, 0.1mm (0.004in) thick.
Assay≥99.995% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only