Indium foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)
Indium foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)
Indium foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Indium foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011383.GH
also known as 011383-GH
Price (USD)
791.65
Special offer
Online exclusive
Ends: 30-Jun-2026
931.00
Save 139.35 (15%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
791.65
Special offer
Online exclusive
Ends: 30-Jun-2026
931.00
Save 139.35 (15%)
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Total Metal Impurities≤0.002%
FormFoil, approx. 1 mm thick
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only