Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis)
Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis)
Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis)
Thermo Scientific Chemicals

Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
100 x 200 mm
Catalog number 011384.GK
also known as 011384-GK
Price (USD)
1,099.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,294.00
Save 194.35 (15%)
Each
Quantity:
100 x 200 mm
Request bulk or custom format
Price (USD)
1,099.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,294.00
Save 194.35 (15%)
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.5 mm thick
Total Metal Impurities≤0.0025% (total metal impurities)
Total Metal ImpuritiesList metal impurities
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only