Indium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis)
Indium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis)
Indium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis)
Thermo Scientific Chemicals

Indium foil, 0.25mm (0.01in) thick, Puratronic™, 99.9975% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 011385.FI
also known as 011385-FI
Price (USD)
192.00
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
192.00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-74-6
ChEBICHEBI:30430
Chemical Name or MaterialIndium foil
EINECS Number231-180-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only