Gold foil, 1.0mm (0.04in) thick, Premion™, 99.9985% (metals basis)
Gold foil, 1.0mm (0.04in) thick, Premion™, 99.9985% (metals basis)
Gold foil, 1.0mm (0.04in) thick, Premion™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Gold foil, 1.0mm (0.04in) thick, Premion™, 99.9985% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Catalog NumberQuantity
011388.FF
also known as 011388-FF
25 x 25 mm
Catalog number 011388.FF
also known as 011388-FF
Price (USD)
3,169.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
3,169.00
Each
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 1.0mm (0.04in) thick.
Appearance (Color)Golden
Assay from Supplier's CofA≥99.9985% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only