Silver(I) oxide, 99+% (metals basis)
Silver(I) oxide, 99+% (metals basis)
Silver(I) oxide, 99+% (metals basis)
Thermo Scientific Chemicals

Silver(I) oxide, 99+% (metals basis)

CAS: 20667-12-3 | Ag2O | 231.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number 011407.14
also known as 011407-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS20667-12-3
IUPAC Name(argentiooxy)silver
Molecular FormulaAg2O
InChI KeyKHJDQHIZCZTCAE-UHFFFAOYSA-N
SMILES[Ag]O[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray to black
FormPowder
Assay (unspecified)Carbonate: Reported
Assay (unspecified)Chloride: Reported
Assay (unspecified)Silver Oxide: Reported
View more
Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only