Potassium perrhenate, 99% (metals basis), Re 64%
Potassium perrhenate, 99% (metals basis), Re 64%
Potassium perrhenate, 99% (metals basis), Re 64%
Thermo Scientific Chemicals

Potassium perrhenate, 99% (metals basis), Re 64%

A powerful oxidizer | CAS: 10466-65-6 | KO4Re | 289.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011411.04
also known as 011411-04
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.70
Save 11.05 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.70
Save 11.05 (15%)
Each
Chemical Identifiers
CAS10466-65-6
IUPAC Namepotassium rheniumoylolate
Molecular FormulaKO4Re
InChI KeyQFKRWIFGDGKWLM-UHFFFAOYSA-N
SMILES[K+].[O-][Re](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max., Re 63.7-64.7%
A powerful oxidizer. Also used for spectrometry, tungsten wire cathode coating color film and as catalyst. Potassium perrhenate is used as strong oxidizing agent and used in the synthesis of technetium and rhenium organometallic radiopharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only