Sodium perrhenate, 99+% (metals basis), Re 68%
Sodium perrhenate, 99+% (metals basis), Re 68%
Sodium perrhenate, 99+% (metals basis), Re 68%
Thermo Scientific Chemicals

Sodium perrhenate, 99+% (metals basis), Re 68%

CAS: 13472-33-8 | NaO4Re | 273.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011412.18
also known as 011412-18
Price (USD)
1,555.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,830.00
Save 274.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
1,555.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,830.00
Save 274.35 (15%)
Each
Chemical Identifiers
CAS13472-33-8
IUPAC Namesodium rheniumoylolate
Molecular FormulaNaO4Re
InChI KeyKMISVFTVBOPTAI-UHFFFAOYSA-N
SMILES[Na+].[O-][Re](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max., Re 68%
Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only