Tantalum rod, 4.5mm (0.18 in.) dia., Puratronic™, 99.96% (metals basis), Nb nominal 50ppm, Thermo Scientific Chemicals
Tantalum rod, 4.5mm (0.18 in.) dia., Puratronic™, 99.96% (metals basis), Nb nominal 50ppm, Thermo Scientific Chemicals
Tantalum rod, 4.5mm (0.18 in.) dia., Puratronic™, 99.96% (metals basis), Nb nominal 50ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum rod, 4.5mm (0.18 in.) dia., Puratronic™, 99.96% (metals basis), Nb nominal 50ppm, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
011441.DM
also known as 011441-DM
100 mm
Catalog number 011441.DM
also known as 011441-DM
Price (USD)
231.65
Special offer
Online exclusive
Ends: 30-Jun-2026
272.00
Save 40.35 (15%)
Each
Quantity:
100 mm
Request bulk or custom format
Price (USD)
231.65
Special offer
Online exclusive
Ends: 30-Jun-2026
272.00
Save 40.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-25-7
ChEBICHEBI:33348
Chemical Name or MaterialTantalum rod
EINECS Number231-135-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only