Copper rod, 7mm (0.3in) dia, Puratronic™, 99.999% (metals basis)
Copper rod, 7mm (0.3in) dia, Puratronic™, 99.999% (metals basis)
Copper rod, 7mm (0.3in) dia, Puratronic™, 99.999% (metals basis)
Copper rod, 7mm (0.3in) dia, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper rod, 7mm (0.3in) dia, Puratronic™, 99.999% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
150 mm
5 x 150 mm
Catalog number 011443.HT
also known as 011443-HT
Price (USD)
945.65
Online Exclusive
1,050.00
Save 104.35 (10%)
Each
Add to cart
Quantity:
5 x 150 mm
Request bulk or custom format
Price (USD)
945.65
Online Exclusive
1,050.00
Save 104.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormRod, 7mm (0.3in) diameter.
Assay from Supplier's CofA≥99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only