Iron foil, 2.0mm (0.08in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 2.0mm (0.08in) thick, Puratronic™, 99.995% (metals basis)
Iron foil, 2.0mm (0.08in) thick, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Iron foil, 2.0mm (0.08in) thick, Puratronic™, 99.995% (metals basis)

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 011447.FF
also known as 011447-FF
Price (USD)
269.65
Online Exclusive
299.00
Save 29.35 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
269.65
Online Exclusive
299.00
Save 29.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7439-89-6
ChEBICHEBI:82664
Chemical Name or MaterialIron foil
EINECS Number231-096-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only