Nickel rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
Nickel rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
Nickel rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
Thermo Scientific Chemicals

Nickel rod, 6.35mm (0.25in) dia, 99.5% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 cm
120 cm
Catalog number 011454.BV
also known as 011454-BV
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Quantity:
30 cm
Request bulk or custom format
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.5% (metals basis)
FormRod, 6.35mm (0.25in) diameter.
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only