Cobalt foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Cobalt foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Cobalt foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Cobalt foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011455.FF
also known as 011455-FF
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
96.00
Save 14.35 (15%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 30-Jun-2026
96.00
Save 14.35 (15%)
Each
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue silver
FormFoil, 0.1mm (0.004in) thick.
Assay from Supplier's CofA≥99.995% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only