Tin foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)
Tin foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)
Tin foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)
Tin foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Tin foil, 1.0mm (0.04in) thick, Puratronic™, 99.9985% (metals basis)

Form: Approx. 9.1g/25 x 50mm | CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 50 mm
50 x 100 mm
100 x 250 mm
Catalog number 011457.FG
also known as 011457-FG
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.20
Save 12.55
Each
Quantity:
25 x 50 mm
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.20
Save 12.55
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9985% (metals basis)
Appearance (Color)Silvery
FormFoil, 1.0mm (0.04in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only