Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals

Silver wire, 1.0mm (0.04in) dia, Premion™, 99.999% (metals basis)

Form: Approx. 8.24g/m | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
250 cm
1000 cm
Catalog number 011466.BW
also known as 011466-BW
Price (USD)
-
Quantity:
50 cm
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSilver
Total Metal Impurities≤ 0.001%
Diameter0.9-1.1mm
FormWire
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only