Silver wire, 0.25mm (0.01in) dia, Premion™, 99.9985% (metals basis)
Silver wire, 0.25mm (0.01in) dia, Premion™, 99.9985% (metals basis)
Silver wire, 0.25mm (0.01in) dia, Premion™, 99.9985% (metals basis)
Silver wire, 0.25mm (0.01in) dia, Premion™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Silver wire, 0.25mm (0.01in) dia, Premion™, 99.9985% (metals basis)

Form: approx. 0.51g/m | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
25 m
Catalog number 011468.G9
also known as 011468-G9
Price (USD)
819.00
Each
Add to cart
Quantity:
25 m
Request bulk or custom format
Price (USD)
819.00
Each
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥99.9985% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only