Tin rod, 6mm (0.2in) dia, Puratronic™, 99.9985% (metals basis)
Tin rod, 6mm (0.2in) dia, Puratronic™, 99.9985% (metals basis)
Tin rod, 6mm (0.2in) dia, Puratronic™, 99.9985% (metals basis)
Thermo Scientific Chemicals

Tin rod, 6mm (0.2in) dia, Puratronic™, 99.9985% (metals basis)

Form: Approx. 2.06g/cm | CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
160 mm
5 x 160 mm
Catalog number 011472.DP
also known as 011472-DP
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Quantity:
160 mm
Request bulk or custom format
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 6mm (0.2in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA>99.9985% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only