Iron(II) iodide, anhydrous, 97% (metals basis)
Iron(II) iodide, anhydrous, 97% (metals basis)
Iron(II) iodide, anhydrous, 97% (metals basis)
Thermo Scientific Chemicals

Iron(II) iodide, anhydrous, 97% (metals basis)

CAS: 7783-86-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 011485.09
also known as 011485-09
Price (USD)
281.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
281.00
Each
Chemical Identifiers
CAS7783-86-0
IUPAC Nameλ²-iron(2+) diiodide
Molecular FormulaFeI2
InChI KeyBQZGVMWPHXIKEQ-UHFFFAOYSA-L
SMILES[Fe++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities3% max.
Iron(II) iodide is used as a catalyst in organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only