Iron(II) fluoride, anhydrous, 98%
Iron(II) fluoride, anhydrous, 98%
Iron(II) fluoride, anhydrous, 98%
Iron(II) fluoride, anhydrous, 98%
Thermo Scientific Chemicals

Iron(II) fluoride, anhydrous, 98%

CAS: 7789-28-8 | F2Fe | 93.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011486.09
also known as 011486-09
Price (USD)
89.40
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
89.40
Each
Add to cart
Chemical Identifiers
CAS7789-28-8
IUPAC Nameλ²-iron(2+) difluoride
Molecular FormulaF2Fe
InChI KeyFZGIHSNZYGFUGM-UHFFFAOYSA-L
SMILES[F-].[F-].[Fe++]
View more
SpecificationsSpecification SheetSpecification Sheet
Formcrystalline solid
Appearance (Color)white
Assay (unspecified)97.6% min
Iron(II) fluoride is used for proteomics research applications. It is also a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only