Aluminum fluoride trihydrate, 97%
Aluminum fluoride trihydrate, 97%
Aluminum fluoride trihydrate, 97%
Thermo Scientific Chemicals

Aluminum fluoride trihydrate, 97%

CAS: 15098-87-0 | AlF3H6O3 | 138.022 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 011496.22
also known as 011496-22
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.10
Save 7.45 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
44.65
Special offer
Online exclusive
Ends: 30-Jun-2026
52.10
Save 7.45 (14%)
Each
Chemical Identifiers
CAS15098-87-0
IUPAC Namealuminium(3+) trihydrate trifluoride
Molecular FormulaAlF3H6O3
InChI KeyBKLZIAYVINRQEJ-UHFFFAOYSA-K
SMILESO.O.O.[F-].[F-].[F-].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only