Citric acid monohydrate, 99.5-100.5% (by anhydrous basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Citric acid monohydrate, 99.5-100.5% (by anhydrous basis), granular, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
50 kg
Catalog number 0115-05
Price (MXN)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
This product is GMP Manufactured and meets J.P., U.S.P, B.P and E.P specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS5949-29-1
Chemical Name or MaterialCitric acid monohydrate
ColorWhite
Melting Point135°C to 152°C
Molecular FormulaC6H8O7·H2O
PackagingPoly bottle, tamper-evident cap
SynonymHydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
GradeMulti-Compendial
Molecular Weight (g/mol)210.14
Percent Purity99.5 to 100.5%
Quantity2.5 kg
Unit SizeEach