Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.99% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals

Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.99% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
011508.FF
also known as 011508-FF
25 x 25 mm
Catalog number 011508.FF
also known as 011508-FF
Price (USD)
1,204.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
1,204.00
Each
Specifications
AppearanceSilvery white foil
Assay Percent Range99.99% (Metals basis)
CAS6-4-7440
ChEBICHEBI:33400
Chemical Name or MaterialPlatinum
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only