Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)
Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)
Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Palladium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)

Form: Approx.0.75g/25 x 25mm | CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011515.GH
also known as 011515-GH
Price (USD)
3,054.65
Special offer
Online exclusive
Ends: 30-Jun-2026
3,594.00
Save 539.35 (15%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
3,054.65
Special offer
Online exclusive
Ends: 30-Jun-2026
3,594.00
Save 539.35 (15%)
Each
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver gray
Assay from Supplier's CofA≥99.9% (metals basis)
FormFoil, 0.1mm (0.004in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only