Palladium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)
Palladium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)
Palladium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Palladium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)

Form: Approx.0.2g/25 x 25mm | CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 011517.FF
also known as 011517-FF
Price (USD)
106.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
106.00
Each
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMaterial is ≥99.9% metals basis and 0.025mm (0.001 in) thick
Certificate of ConformanceCertificate of Conformance Provided
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only