Gold foil, 0.025mm (0.001in) thick, 99.95% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.95% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.95% (metals basis)
Gold foil, 0.025mm (0.001in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Gold foil, 0.025mm (0.001in) thick, 99.95% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 011519.FF
also known as 011519-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Appearance (Color)Gold
Comment0.022 - 0.028mm thick
FormFoil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only