Lithium hexafluorophosphate, 98%
Lithium hexafluorophosphate, 98%
Lithium hexafluorophosphate, 98%
Lithium hexafluorophosphate, 98%
Thermo Scientific Chemicals

Lithium hexafluorophosphate, 98%

Used As An Electrolyte in Li-Ion Batteries | CAS: 21324-40-3 | F6LiP | 151.904 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 011529.09
also known as 011529-09
Price (USD)
82.90
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
82.90
Each
Chemical Identifiers
CAS21324-40-3
IUPAC Namelithium(1+) hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6LiP
InChI KeyAXPLOJNSKRXQPA-UHFFFAOYSA-N
SMILES[Li+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (unspecified)≥97.5%

Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only