Potassium hexafluorophosphate, 95% min
Potassium hexafluorophosphate, 95% min
Potassium hexafluorophosphate, 95% min
Thermo Scientific Chemicals

Potassium hexafluorophosphate, 95% min

CAS: 17084-13-8 | F6KP | 184.062 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 011531.30
also known as 011531-30
Price (USD)
87.65
Online Exclusive
97.50
Save 9.85 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
97.50
Save 9.85 (10%)
Each
Chemical Identifiers
CAS17084-13-8
IUPAC Namepotassium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6KP
InChI KeyYZDGRYDIGCWVND-UHFFFAOYSA-N
SMILES[K+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)94.1% min
Potassium hexafluorophosphate is a soluble neutral salt used for synthesis. It is a source of the hexafluorophosphate anion, a non-coordinating anion that confers lipophilicity to its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only