Titanium(IV) fluoride, 98%
Titanium(IV) fluoride, 98%
Titanium(IV) fluoride, 98%
Titanium(IV) fluoride, 98%
Thermo Scientific Chemicals

Titanium(IV) fluoride, 98%

CAS: 7783-63-3 | TiF4
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011540.14
also known as 011540-14
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
101.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 30-Jun-2026
101.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7783-63-3
IUPAC Nametitanium(4+) tetrafluoride
Molecular FormulaF4Ti
InChI KeyXROWMBWRMNHXMF-UHFFFAOYSA-J
SMILES[F-].[F-].[F-].[F-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
FormCrystalline
Appearance (Color)White
Titanium(IV) fluoride is used for preparation of glycosyl fluorides, glycosylation, preparation of fluorohydrins, addition of carbanions to aldehydes and imines. It is a catalyst for chemoselective synthesis and deprotection of geminal diacetates of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only