Zinc fluoride, anhydrous, 99% min
Zinc fluoride, anhydrous, 99% min
Zinc fluoride, anhydrous, 99% min
Thermo Scientific Chemicals

Zinc fluoride, anhydrous, 99% min

CAS: 7783-49-5 | F2Zn | 103.377 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 011541.A1
also known as 011541-A1
Price (USD)
976.65
Online Exclusive
1,085.00
Save 108.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
976.65
Online Exclusive
1,085.00
Save 108.35 (10%)
Each
Chemical Identifiers
CAS7783-49-5
IUPAC Namezinc(2+) difluoride
Molecular FormulaF2Zn
InChI KeyBHHYHSUAOQUXJK-UHFFFAOYSA-L
SMILES[F-].[F-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99% min
It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only