Chromium carbide, 99.5% (metals basis)
Chromium carbide, 99.5% (metals basis)
Chromium carbide, 99.5% (metals basis)
Thermo Scientific Chemicals

Chromium carbide, 99.5% (metals basis)

CAS: 12012-35-0 | C2Cr3 | 180.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 011547.18
also known as 011547-18
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.50
Save 11.85
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.50
Save 11.85
Each
Chemical Identifiers
CAS12012-35-0
IUPAC Name{[(chromiomethyl)chromio]methylidyne}chromium
Molecular FormulaC2Cr3
InChI KeyUFGZSIPAQKLCGR-UHFFFAOYSA-N
SMILES[Cr]#C[Cr]C#[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Chromium carbide is useful in the surface treatment of metal components. It is used to coat the surface of another metal in a technique known as thermal spraying. It is also used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only