Tin(II) oxide, 99.9% (metals basis)
Tin(II) oxide, 99.9% (metals basis)
Tin(II) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals
Tin(II) oxide, 99.9% (metals basis)
Reducing agent, soft abrasive, preparation of stannous salts | CAS: 21651-19-4 | OSn | 134.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 011569.14
also known as 011569-14
Price (USD)
40.30
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
40.30
Each
Chemical Identifiers
CAS21651-19-4
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
It is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.