Rhodium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
Rhodium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
Rhodium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Rhodium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)

CAS: 7440-16-6 | Rh | 102.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
Catalog number 011578.BU
also known as 011578-BU
Price (USD)
-
Quantity:
25 cm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-16-6
ChEBICHEBI:33359
Chemical Name or MaterialRhodium wire
EINECS Number231-125-0
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only