Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals

Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%

CAS: 10049-21-5 | H2NaO4P x H2O | 137.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 011591.30
also known as 011591-30
Price (USD)
55.65
Online Exclusive
61.70
Save 6.05 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa: ≤0.005%
Impurity contentCl: ≤5ppm
Impurity contentFe: ≤0.001%
Impurity contentK: ≤0.01%
Impurity contentSO₄:<0.003%
View more
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only