Magnesium sulfate heptahydrate, ACS, 98.0-102.0%
Magnesium sulfate heptahydrate, ACS, 98.0-102.0%
Magnesium sulfate heptahydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, ACS, 98.0-102.0%

For Fireproofing, Textiles, Catalyst Carrier | CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 011596.A3
also known as 011596-A3
Price (USD)
130.65
Online exclusive
145.00
Save 14.35
Each
Quantity:
2 kg
Request bulk or custom format
Price (USD)
130.65
Online exclusive
145.00
Save 14.35
Each
Chemical Identifiers
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0-102.0% MgSO₄?7H₂O
Impurity contentNH₄ 0.002%
Impurity contentNO₃ 0.002%
Impurity contentSr 0.005%
pHof a 5% solution 5.0-8.2 at 25?
View more
Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only