Silver(I) fluoride, 98%
Silver(I) fluoride, 98%
Silver(I) fluoride, 98%
Thermo Scientific Chemicals

Silver(I) fluoride, 98%

Light and Moisture Sensitive and Hygroscopic | CAS: 7775-41-9 | AgF | 126.867 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 011609.14
also known as 011609-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7775-41-9
IUPAC Namesilver(1+) fluoride
Molecular FormulaAgF
InChI KeyREYHXKZHIMGNSE-UHFFFAOYSA-M
SMILES[F-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Silver(I) fluoride is used as a fluorination and desilylation reagent in organic synthesis. In medical filed, it is used as topical caries in aqueous form in dentistry. It is employed in organofluorine chemistry for addition of fluoride across multiple bonds, especially to prepare perfluoroalkylsilver(I) by reacting with alkenes. Further, it is involved in the preparation of tetralkylammonium fluorides by the reaction with tetralkylammonium bromide. In addition to this, it is used as a desulfuration-fluorination reagent on thiourea derived substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only