Lithium perchlorate trihydrate, Reagent Grade
Lithium perchlorate trihydrate, Reagent Grade
Lithium perchlorate trihydrate, Reagent Grade
Lithium perchlorate trihydrate, Reagent Grade
Thermo Scientific Chemicals

Lithium perchlorate trihydrate, Reagent Grade

CAS: 13453-78-6 | ClH6LiO7 | 160.431 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 011638.A1
also known as 011638-A1
Price (USD)
228.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
228.00
Each
Add to cart
Chemical Identifiers
CAS13453-78-6
IUPAC Namelithium(1+) trihydrate perchlorate
Molecular FormulaClH6LiO7
InChI KeyJAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES[Li+].O.O.O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)63.0-68.0% LiClO₄
It is used as an oxygen source for chemical oxygen generators, as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte, and act as chaotropic agent to denature proteins. It is used in Baylis-Hillman reaction as a catalyst in the coupling of alpha,beta-unsaturated carbonyl compounds with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only