Cobalt(II) perchlorate hexahydrate, Reagent Grade
Cobalt(II) perchlorate hexahydrate, Reagent Grade
Cobalt(II) perchlorate hexahydrate, Reagent Grade
Thermo Scientific Chemicals

Cobalt(II) perchlorate hexahydrate, Reagent Grade

CAS: 13478-33-6 | Cl2CoO8 | 257.825 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 011652.09
also known as 011652-09
Price (USD)
31.10
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
31.10
Each
Chemical Identifiers
CAS13478-33-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Co 15.5-16.7%
Cobalt(II) perchlorate hexahydrate is used as chemical organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only