Silver acetate, anhydrous, 99%
Silver acetate, anhydrous, 99%
Silver acetate, anhydrous, 99%
Thermo Scientific Chemicals

Silver acetate, anhydrous, 99%

CAS: 563-63-3 | C2H3AgO2 | 166.912 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 011660.14
also known as 011660-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS563-63-3
IUPAC Namesilver(1+) acetate
Molecular FormulaC2H3AgO2
InChI KeyCQLFBEKRDQMJLZ-UHFFFAOYSA-M
SMILES[Ag+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormMicrocrystalline powder or powder
Assay from Supplier's CofA≥98.5% to ≤101.5% [UK sourced material]
Assay from Supplier's CofA64.5 ± 0.5% Ag w/w [US sourced material]
CommentMaterial sourced either in UK or USA
View more
It is a reagent in the laboratory as a source of silver ions lacking an oxidizing anion. It is a reagent for direct ortho-arylation, and for conversion of organohalogen compounds into alcohols. Woodward cis-hydroxylation reaction employs silver acetate and iodine for selective conversin of alkenes into cis-diols. Silver acetate is the more preferred reagent for facile carbonylation of primary and secondary amines. It is also employed in the preparation of highly reflective, conductive silvered polymer films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only