Antimony(III) sulfide, 99.999% (metals basis)
Antimony(III) sulfide, 99.999% (metals basis)
Antimony(III) sulfide, 99.999% (metals basis)
Thermo Scientific™

Antimony(III) sulfide, 99.999% (metals basis)

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011683.09
also known as 011683-09
Price (USD)
103.65
Online Exclusive
115.00
Save 11.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
103.65
Online Exclusive
115.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1345-04-6
IUPAC Namediantimony(3+) trisulfanediide
Molecular FormulaS3Sb2
InChI KeyNVWBARWTDVQPJD-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[Sb+3].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used in glitter compositions, fountain compositions, flash powder and vulcanizing rubber. It is employed in the production of ruby-colored glass and in plastics, as a fire retardant, in explosives, matches, and fireworks. It can be used to prepare a series of thioantimonous salts and Schlippe's salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only