Ruthenium(III) chloride oxide, ammoniated
Ruthenium(III) chloride oxide, ammoniated
Ruthenium(III) chloride oxide, ammoniated
Thermo Scientific Chemicals

Ruthenium(III) chloride oxide, ammoniated

Microscopic stain | CAS: 11103-72-3 | Cl6H46N14O2Ru3 | 790.374 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 011806.03
also known as 011806-03
Price (USD)
109.65
Special offer
Online exclusive
Ends: 28-Dec-2026
129.00
Save 19.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 28-Dec-2026
129.00
Save 19.35
Each
Chemical Identifiers
CAS11103-72-3
IUPAC Nametris(λ²-ruthenium(2+)) tetradecaamine dihydrate hexachloride
Molecular FormulaCl6H46N14O2Ru3
InChI KeyJQJSTVUROJELSR-UHFFFAOYSA-H
SMILESN.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[Ru++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.1%
Appearance (Color)Red to red-brown
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥34.0% Ruthenium
Microscopic stainRuthenium(III) chloride oxide, ammoniated is used as a reagent on plant material for staining pectin, gum, animal tissues and bacteria. It is also used in histology to stain aldehyde fixed mucopolysaccharides. Further, it serves as a pharmacological tool to study specific cellular mechanisms. It serves as an inhibitor of mitochondrial calcium uniporter. In addition, it is involved in reducing airway inflammation arises by pepper spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only