12-Tungstophosphate hydrate, Reagent Grade
12-Tungstophosphate hydrate, Reagent Grade
12-Tungstophosphate hydrate, Reagent Grade
12-Tungstophosphate hydrate, Reagent Grade
Thermo Scientific Chemicals

12-Tungstophosphate hydrate, Reagent Grade

CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 011827.30
also known as 011827-30
Price (USD)
367.65
Online Exclusive
408.00
Save 40.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
367.65
Online Exclusive
408.00
Save 40.35 (10%)
Each
Chemical Identifiers
CAS12501-23-4
IUPAC Name12-Tungstophosphate hydrate
Molecular Weight (g/mol)2880.20
MDL NumberMFCD00011342
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to yellow
Impurity contentAmmonium: ≤0.005%
Impurity contentIron: ≤0.002%
Impurity contentNitrate: ≤0.010%
Impurity contentSulfate: ≤0.020%
View more
12-Tungstophosphate hydrate is used in histology as a component for staining of cell specimens. It binds to fibrin, collagen and fibers of connective tissues. It is also used to replace the anions of dyes from these materials. In solution, it acts as a homogeneous catalyst and involved in the hydrolysis of propene to give 2-propanol. It is supported with alumina and silica and act as a heterogeneous catalyst involved in the dehydration of 2-propanol to propene and methanol to hydrocarbons. In analytical chemistry, it is used to precipitate proteins and termed as universal precipitant for polar proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only