Indium(III) chloride, anhydrous, 99.999% (metals basis)
Indium(III) chloride, anhydrous, 99.999% (metals basis)
Indium(III) chloride, anhydrous, 99.999% (metals basis)
Thermo Scientific Chemicals

Indium(III) chloride, anhydrous, 99.999% (metals basis)

CAS: 10025-82-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
250 g
Catalog number 011856.09
also known as 011856-09
Price (USD)
157.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
157.00
Each
Chemical Identifiers
CAS10025-82-8
IUPAC Nameindium(3+) trichloride
Molecular FormulaCl3In
InChI KeyPSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystals or crystalline powder
Assay from Suppliers CofAReport Metals Analysis
Total Metal Impurities≤0.001%
It is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only