Cadmium chloride, anhydrous, 99.99% (metals basis)
Cadmium chloride, anhydrous, 99.99% (metals basis)
Cadmium chloride, anhydrous, 99.99% (metals basis)
Thermo Scientific Chemicals

Cadmium chloride, anhydrous, 99.99% (metals basis)

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011860.14
also known as 011860-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS10108-64-2
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Cadmium chloride is a precursor material for cadmium sulfide, which is a brilliant yellow pigment known as Cadmium Yellow. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium. It is a mild Lewis acid, and can form tetrachlorocadmium species. It is used in the preparation of organocadmium compounds, such as dialkyl or diaryl cadmium, which are useful for the synthesis of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only