Cadmium sulfate, anhydrous, ACS, 99+%
Cadmium sulfate, anhydrous, ACS, 99+%
Cadmium sulfate, anhydrous, ACS, 99+%
Thermo Scientific Chemicals

Cadmium sulfate, anhydrous, ACS, 99+%

CAS: 10124-36-4 | CdO4S | 208.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011861.22
also known as 011861-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS10124-36-4
IUPAC Namecadmium(2+) sulfate
Molecular FormulaCdO4S
InChI KeyQCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES[Cd++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.0% CdSO₄ min
Impurity contentCa 0.01%
Impurity contentCl 0.001%
Impurity contentCu 0.002%
Impurity contentPb 0.003%
View more
Cadmium sulfate is used in pigments and electroplating. It acts as an accelerator in cement formation. It is also employed as lubricants. Further, it is useful for the electroplating of cadmium in electronic circuits. It serves as a precursor to prepare cadmium-based pigment like cadmium sulfide. In the Western standard cell, it is used for electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only